Learn More
Waters Corp XSelect Premier Peptide CSH C18 Column, 130 Å, 2.5 μm, 2.1 x 100 mm
Catalog No. 502392025 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This column is specifically designed for peptide separations, featuring a low and carefully-defined concentration of positive charges that ensures excellent peak shape. It is ideal for LC and LC-MS applications due to minimal dependence on strong ion pairing agents. The 2.1 mm ID columns are recommended for low dispersion systems and are part of the MaxPeak Premier Class, delivering enhanced reproducibility, peak shape, and recovery for metal-sensitive analytes.
  • First choice for peptide separations.
  • Ensures excellent peak shape for peptides using FA- or TFA-containing mobile phases.
  • Quality controlled with cytochrome c digest standards.
  • Unique design includes an integrated, replaceable 5 mm pre-column to extend analytical column life.
  • VanGuard FIT cartridge replacements use the same qualified sorbent.
  • Suggested for peptide separations, analysis, mapping, and synthetic peptides.
Catalog No. 50-239-2025 Supplier Waters Corp Supplier No. 186010713
This column is specifically designed for peptide separations, featuring a low and carefully-defined concentration of positive charges that ensures excellent peak shape. It is ideal for LC and LC-MS applications due to minimal dependence on strong ion pairing agents. The 2.1 mm ID columns are recommended for low dispersion systems and are part of the MaxPeak Premier Class, delivering enhanced reproducibility, peak shape, and recovery for metal-sensitive analytes.
  • First choice for peptide separations.
  • Ensures excellent peak shape for peptides using FA- or TFA-containing mobile phases.
  • Quality controlled with cytochrome c digest standards.
  • Unique design includes an integrated, replaceable 5 mm pre-column to extend analytical column life.
  • VanGuard FIT cartridge replacements use the same qualified sorbent.
  • Suggested for peptide separations, analysis, mapping, and synthetic peptides.