Learn More
Waters Corp UHPLC Column, 100 Å Pore, 2.5 μm Particle, 2.1 mm ID, 100 mm Length
Supplier: Waters Corp 186010725
The XSelect Premier Peptide HSS T3 Columns are engineered for separations demanding silica-based selectivities or enhanced retention of hydrophilic peptides. These MaxPeak Premier columns offer superior chromatographic performance, boosting reproducibility, refining peak shape, and improving recovery for metal-sensitive analytes. The design includes a fully integrated, replaceable 5 mm pre-column, extending analytical column lifespan without compromising resolution. VanGuard FIT Cartridge replacements contain the same qualified sorbent and are easily replaceable.
- Carbon load: 11.0%.
- Endcapping: Yes.
- Inner diameter: 2.1 mm.
- Mode: Reversed Phase (RP).
- Particle size: 2.5 μm.
- pH range: 2 to 8.
- Pore size: 100 Å.
- USP number: L1.
- Length: 100 mm.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.