Learn More
Waters Corp XSelect Premier Peptide HSS T3 Column, 100 Å, 2.5 μm, 4.6 x 150 mm
Catalog No. 502392738 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This XSelect Premier Peptide HSS T3 Column with MaxPeak High Performance Surfaces is designed for peptide analysis. It minimizes detrimental analyte/surface interactions, enhancing recovery, sensitivity, and reproducibility for metal-sensitive analytes like acidic peptides. Utilizing HSS T3 particle technology, it offers increased retentivity and mechanical stability, ideal for separating small, polar peptides.
  • Minimizes loss of metal-sensitive analytes, including acidic peptides, by reducing analyte/surface interactions.
  • Ideal for separating small, polar peptides, offering greater retentivity.
  • Provides increased retentivity and mechanical stability.
  • Offers reduced conditioning, improved sensitivity, enhanced recovery, and simpler mobile phases.
Catalog No. 50-239-2738 Supplier Waters Corp Supplier No. 186010729
This XSelect Premier Peptide HSS T3 Column with MaxPeak High Performance Surfaces is designed for peptide analysis. It minimizes detrimental analyte/surface interactions, enhancing recovery, sensitivity, and reproducibility for metal-sensitive analytes like acidic peptides. Utilizing HSS T3 particle technology, it offers increased retentivity and mechanical stability, ideal for separating small, polar peptides.
  • Minimizes loss of metal-sensitive analytes, including acidic peptides, by reducing analyte/surface interactions.
  • Ideal for separating small, polar peptides, offering greater retentivity.
  • Provides increased retentivity and mechanical stability.
  • Offers reduced conditioning, improved sensitivity, enhanced recovery, and simpler mobile phases.