Learn More
Waters Corp XSelect Column, 2.5 μm, 2.1 mm × 50 mm, 100 Å
Catalog No. 502391372 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These columns are engineered for silica-based separations, ideal for retaining hydrophilic peptides. Each batch undergoes stringent QC testing with a tryptic digest of cytochrome c. Part of the MaxPeak Premier class, these columns boost reproducibility, peak shape, and recovery for metal-sensitive analytes. They feature a fully integrated, replaceable 5 mm pre-column to prolong analytical column life.
  • Ideal for silica-based separations and hydrophilic peptide retention.
  • Stringently QC tested for consistent performance.
  • 2.1 mm ID columns suit low dispersion UPLC/UHPLC and LC-MS methods.
  • MaxPeak Premier class boosts reproducibility and recovery for metal-sensitive analytes.
  • Integrated 5 mm pre-column extends life without impacting resolution; easily replaceable.
  • VanGuard FIT Cartridge replacements use qualified sorbent.
  • Recommended for peptide separations and analysis.
Catalog No. 50-239-1372 Supplier Waters Corp Supplier No. 186010724
These columns are engineered for silica-based separations, ideal for retaining hydrophilic peptides. Each batch undergoes stringent QC testing with a tryptic digest of cytochrome c. Part of the MaxPeak Premier class, these columns boost reproducibility, peak shape, and recovery for metal-sensitive analytes. They feature a fully integrated, replaceable 5 mm pre-column to prolong analytical column life.
  • Ideal for silica-based separations and hydrophilic peptide retention.
  • Stringently QC tested for consistent performance.
  • 2.1 mm ID columns suit low dispersion UPLC/UHPLC and LC-MS methods.
  • MaxPeak Premier class boosts reproducibility and recovery for metal-sensitive analytes.
  • Integrated 5 mm pre-column extends life without impacting resolution; easily replaceable.
  • VanGuard FIT Cartridge replacements use qualified sorbent.
  • Recommended for peptide separations and analysis.