Learn More
Waters Corp Peptide Column, XSelect Premier HSS T3, 100Å, 2.5μm
Catalog No. 502392719 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The XSelect Premier Peptide HSS T3 Column features 100 Å pore size, 2.5 μm particle size, and 4.6 x 50 mm dimensions, and is designed for improved peptide analysis.
  • Features MaxPeak High Performance Surfaces
  • Includes HSS Technology
Catalog No. 50-239-2719 Supplier Waters Corp Supplier No. 186010727
The XSelect Premier Peptide HSS T3 Column features 100 Å pore size, 2.5 μm particle size, and 4.6 x 50 mm dimensions, and is designed for improved peptide analysis.
  • Features MaxPeak High Performance Surfaces
  • Includes HSS Technology