Learn More
Pyramex™ XSG™ Ballistic Lens Kit
Catalog No. 19155339 Shop All Pyramex Safety Products Products
Change view
Click to view available options
Lens Coating:
Anti-fog
Catalog No. Lens Coating
19-155-339 Anti-fog
1 options
This item is not returnable. View return policy
Catalog No. 19-155-339 Supplier Pyramex™ Supplier No. GB4010KIT
Add to Cart
Add to Cart
This item is not returnable. View return policy
Interchangeable ballistic rated lens kit
  • Clear Ballistic, Gray Ballistic and Amber Ballistic Interchangeable Lenses. All lenses are anti-fog
  • Black Nylon Case for storage
  • Flame resistant foam padding forms a seal to keep out dust and airborne particles
  • Strap has Quick Fit retainer for easy on/off
  • Interchangeable temples/strap included with each pair
  • Passes MIL-PRF 32432 High Velocity Impact Standards
  • Specifications
    Lens Coating Anti-fog
    Lens Type H2X
    Certifications/Compliance ANSI Z87.1
    Lens Tint Clear, Gray, Amber
    Product Type Sealed Eyewear
    Frame Color Black
    Includes Clear Ballistic, Gray Ballistic and Amber Ballistic Interchangeable Lenses, Black Nylon Case for storage, Interchangeable temples/strap
    Material Polycarbonate, Nylon
    Disposable Reusable
    Lens Material Polycarbonate
    Frame Material Nylon
    UV Protection U6
    Frame Type Sealed