Learn More
Pyramex™ XSG™ Readers Safety Eyewear
Catalog No. 19155344 Shop All Pyramex Safety Products Products
Change view
Click to view available options
Magnification Power:
+1.50
+2.00
+2.50
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Magnification Power
19-155-344 +2.50
19-155-342 +1.50
19-155-343 +2.00
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 19-155-344 Supplier Pyramex™ Supplier No. GB4010STR25
Add to Cart
Add to Cart
This item is not returnable. View return policy
Includes polyester strap
  • Flame resistant foam padding
  • Strap has quick fit retainer
  • Interchangeable temples and strap included
  • Specifications
    Lens Type H2X
    Certifications/Compliance ANSI Z87.1,CSA Z94.3-07
    Lens Tint Clear
    Frame Color Black
    Magnification Power +2.50
    Weight (Metric) 35 g
    Lens Material Polycarbonate
    Frame Material Nylon