Learn More
Pyramex™ XSG™ Safety Eyewear
Catalog No. 19155346 Shop All Pyramex Safety Products Products
Change view
Click to view available options
Lens Coating:
Anti-fog
Anti-fog, Anti-scratch
Anti-fog, Anti-scratch, Anti-static
Lens Tint:
Clear
Gray
Indoor/Outdoor, Mirror
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Lens Coating Lens Tint
19-155-346 Anti-fog Indoor/Outdoor, Mirror
19-155-340 Anti-fog, Anti-scratch, Anti-static Clear
19-155-341 Anti-fog, Anti-scratch Clear
19-155-345 Anti-fog, Anti-scratch, Anti-static Gray
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 19-155-346 Supplier Pyramex™ Supplier No. GB4080ST
Add to Cart
Add to Cart
This item is not returnable. View return policy
Includes polyester strap
  • Flame resistant foam padding forms a seal to keep out dust and airborne particles
  • Strap has Quick Fit retainer for easy on/off
  • Interchangeable temples/strap included with each pair
  • Specifications
    Lens Coating Anti-fog
    Certifications/Compliance ANSI Z87.1,CSA Z94.3-07
    Certifications/Compliance (Europe) EN-166
    Lens Tint Indoor/Outdoor, Mirror
    Product Type Safety Spectacles
    Frame Color Black
    Weight (Metric) 28 g
    Lens Material Polycarbonate
    Frame Material Nylon