Learn More
Medchemexpress LLC XST-14 | 2607143-50-8 | 99.3% | 291.34 | 50 MG
SDP
Catalog No. 5000350445
Encompass_Preferred
XST-14 is a potent, competitive, and highly selective ULK1 inhibitor with an IC50 of 26.6 nM. It induces autophagy inhibition by reducing the phosphorylation of the ULK1 downstream substrate. XST-14 also induces apoptosis in hepatocellular carcinoma (HCC) cells and exhibits antitumor effects.
  • Potent, competitive, and highly selective ULK1 inhibitor
  • Induces autophagy inhibition
  • Reduces phosphorylation of ULK1 downstream substrate
  • Induces apoptosis in hepatocellular carcinoma (HCC) cells
  • Exhibits antitumor effects
Catalog No. 50-003-50445 Supplier Medchemexpress LLC Supplier No. HY13750650MG
XST-14 is a potent, competitive, and highly selective ULK1 inhibitor with an IC50 of 26.6 nM. It induces autophagy inhibition by reducing the phosphorylation of the ULK1 downstream substrate. XST-14 also induces apoptosis in hepatocellular carcinoma (HCC) cells and exhibits antitumor effects.
  • Potent, competitive, and highly selective ULK1 inhibitor
  • Induces autophagy inhibition
  • Reduces phosphorylation of ULK1 downstream substrate
  • Induces apoptosis in hepatocellular carcinoma (HCC) cells
  • Exhibits antitumor effects