Learn More
Medchemexpress LLC XST-14 | 2607143-50-8 | 99.3% | C16H21NO4 | 10MG
SDP
Catalog No. 5000260442
Encompass_Preferred
XST-14 is a potent, competitive, and highly selective ULK1 inhibitor used in research to modulate autophagy and related signaling pathways. It demonstrates low-nanomolar activity against ULK1 and has documented off-target kinase activity at higher concentrations. The compound is provided as a solid with high purity and solubility data to support in vitro and in vivo formulation.
  • Potent ULK1 inhibition (IC50 = 26.6 nM).
  • High purity (99.3%).
  • Soluble in DMSO (250 mg/mL) for stock preparation.
  • Includes in vivo formulation guidance for dosing studies.
  • Available as small research quantities and ready-to-use DMSO solutions.
Catalog No. 50-002-60442 Supplier Medchemexpress LLC Supplier No. HY13750610MG
XST-14 is a potent, competitive, and highly selective ULK1 inhibitor used in research to modulate autophagy and related signaling pathways. It demonstrates low-nanomolar activity against ULK1 and has documented off-target kinase activity at higher concentrations. The compound is provided as a solid with high purity and solubility data to support in vitro and in vivo formulation.
  • Potent ULK1 inhibition (IC50 = 26.6 nM).
  • High purity (99.3%).
  • Soluble in DMSO (250 mg/mL) for stock preparation.
  • Includes in vivo formulation guidance for dosing studies.
  • Available as small research quantities and ready-to-use DMSO solutions.