Learn More
Medchemexpress LLC xStAx-VHLL TFA | 100.0% | 3231.89 | 5 MG
SDP
Catalog No. 5000399842
Encompass_Preferred
xStAx-VHLL TFA is a β-catenin PROTAC degrader that promotes ubiquitination and proteasome degradation of β-catenin. It inhibits the Wnt/β-catenin signaling pathway. xStAx-VHLL TFA can also inhibit the proliferation of colon cancer cells and demonstrates anti-tumor activity.
  • Induces β-catenin degradation in a dose-dependent manner in colorectal cancer cells in vitro
  • Inhibits cell proliferation in vitro
  • Inhibits Wnt/β-catenin signaling in HEK293T cells in vitro
  • Shows antitumor activity in a mouse intestinal tumor model in vivo
  • Reduces tumor number and active β-catenin protein levels in APC min/+ mice in vivo
Catalog No. 50-003-99842 Supplier Medchemexpress LLC Supplier No. HYP5819A5MG
xStAx-VHLL TFA is a β-catenin PROTAC degrader that promotes ubiquitination and proteasome degradation of β-catenin. It inhibits the Wnt/β-catenin signaling pathway. xStAx-VHLL TFA can also inhibit the proliferation of colon cancer cells and demonstrates anti-tumor activity.
  • Induces β-catenin degradation in a dose-dependent manner in colorectal cancer cells in vitro
  • Inhibits cell proliferation in vitro
  • Inhibits Wnt/β-catenin signaling in HEK293T cells in vitro
  • Shows antitumor activity in a mouse intestinal tumor model in vivo
  • Reduces tumor number and active β-catenin protein levels in APC min/+ mice in vivo