Learn More
Medchemexpress LLC XStAx-VHLL TFA | 99.9% | 3231.89 | 1 MG
SDP
Catalog No. 5000398092
Encompass_Preferred
xStAx-VHLL TFA is a β-catenin PROTAC degrader that promotes ubiquitination and proteasome degradation of β-catenin. It inhibits the Wnt/β-catenin signaling pathway, can inhibit the proliferation of colon cancer cells, and has anti-tumor activity.
  • Promotes ubiquitination and proteasome degradation of β-catenin.
  • Inhibits the Wnt/β-catenin signaling pathway.
  • Inhibits the proliferation of colon cancer cells.
  • Exhibits anti-tumor activity in mouse intestinal tumor models.
  • Induces β-catenin degradation and inhibits cell proliferation in colorectal cancer cells.
  • Inhibits Wnt/β-catenin signaling in HEK293T cells.
Catalog No. 50-003-98092 Supplier Medchemexpress LLC Supplier No. HYP5819A1MG
xStAx-VHLL TFA is a β-catenin PROTAC degrader that promotes ubiquitination and proteasome degradation of β-catenin. It inhibits the Wnt/β-catenin signaling pathway, can inhibit the proliferation of colon cancer cells, and has anti-tumor activity.
  • Promotes ubiquitination and proteasome degradation of β-catenin.
  • Inhibits the Wnt/β-catenin signaling pathway.
  • Inhibits the proliferation of colon cancer cells.
  • Exhibits anti-tumor activity in mouse intestinal tumor models.
  • Induces β-catenin degradation and inhibits cell proliferation in colorectal cancer cells.
  • Inhibits Wnt/β-catenin signaling in HEK293T cells.