Learn More
Pipette Supplies, Inc. XSTREAM LI-ION BATTERY
SDP
Catalog No. NC0920119
Encompass
Eppendorf stream, Xstream Li-Ion Battery. Recharging the battery takes approximately two hours. It is possible to continue working while charging. The implemented Lithium-ion battery has no memory-effect. A fully charged battery allows for approximately 2000 dispensing steps.
Catalog No. NC0920119 Supplier Pipette Supplies, Inc. Supplier No. EP 022462407
Eppendorf stream, Xstream Li-Ion Battery. Recharging the battery takes approximately two hours. It is possible to continue working while charging. The implemented Lithium-ion battery has no memory-effect. A fully charged battery allows for approximately 2000 dispensing steps.