Learn More
WELCH MATERIALS INC Xtimate C18, 10µm, 10×250mm
Catalog No. NC2052723
Encompass_Preferred
Prep Column, Xtimate C18, 10µm, 10×250mm, pH range: 1.0-12.5, Pore Size: 120Å, USP listing: L1, Carbon Loading: 14%, Surface Area: 320m2/g, endcapped.
Catalog No. NC2052723 Supplier WELCH MATERIALS INC Supplier No. 0161031102
Add to Cart
Add to Cart
Prep Column, Xtimate C18, 10µm, 10×250mm, pH range: 1.0-12.5, Pore Size: 120Å, USP listing: L1, Carbon Loading: 14%, Surface Area: 320m2/g, endcapped.