Learn More
WELCH MATERIALS INC Xtimate C18, 10µm, 120Å, 4.6×250mm
Catalog No. NC2052228
Encompass_Preferred
Analytical Column, Xtimate C18, 10μm, 120Å, 4.6×250mm, pH range: 1.0-12.5, Pore Size: 120Å, USP listing: L1, Carbon Loading: 14%, Surface Area: 320m2/g, endcapped.
Catalog No. NC2052228 Supplier WELCH MATERIALS INC Supplier No. 0010131043
Analytical Column, Xtimate C18, 10μm, 120Å, 4.6×250mm, pH range: 1.0-12.5, Pore Size: 120Å, USP listing: L1, Carbon Loading: 14%, Surface Area: 320m2/g, endcapped.