Learn More
WELCH MATERIALS INC Xtimate C18, 40-70µm, 120Å, 1kg
Catalog No. NC2052759
Encompass_Preferred
Bulk Media, Xtimate C18, 40-70μm, pH range: 1.0-12.5, Pore Size: 120Å, USP listing: L1, Carbon Loading: 14%, Surface Area: 320m2/g, endcapped. Packing: 1kg/bottle.
Catalog No. NC2052759 Supplier WELCH MATERIALS INC Supplier No. 0171011100
Add to Cart
Add to Cart
Request a Quote
Bulk Media, Xtimate C18, 40-70μm, pH range: 1.0-12.5, Pore Size: 120Å, USP listing: L1, Carbon Loading: 14%, Surface Area: 320m2/g, endcapped. Packing: 1kg/bottle.