Learn More
WELCH MATERIALS INC Xtimate C18, Cartridge: 5µm,120Å, 4.6×10mm, 10pcs/pk
Catalog No. NC2052535
Encompass_Preferred
Cartridge: Xtimate C18, 5µm,120Å, 4.6×10mm, pH range: 1.0-12.5, Pore Size: 120Å, USP listing: L1, Carbon Loading: 14%, Surface Area: 320m2/g, endcapped. Matched HPLC Column ID: 3.0-8.0, Matched Guard Column Holder: 00808-01101 or 00808-01108, 10pcs/pk.
Catalog No. NC2052535 Supplier WELCH MATERIALS INC Supplier No. 008080410110
Add to Cart
Add to Cart
Cartridge: Xtimate C18, 5µm,120Å, 4.6×10mm, pH range: 1.0-12.5, Pore Size: 120Å, USP listing: L1, Carbon Loading: 14%, Surface Area: 320m2/g, endcapped. Matched HPLC Column ID: 3.0-8.0, Matched Guard Column Holder: 00808-01101 or 00808-01108, 10pcs/pk.