Learn More
WELCH MATERIALS INC Xtimate C8, 10µm, 120Å, 4.6×250mm
Supplier:  WELCH MATERIALS INC 0010231043
Encompass_Preferred
Analytical Column, Xtimate C8, 10μm, 120Å, 4.6×250mm, pH range: 1.0-12.5, Pore Size: 120Å, USP listing: L7, Carbon Loading: 10%, Surface Area: 320m2/g
Catalog No. NC2052230