Learn More
WELCH MATERIALS INC Xtimate C8, Cartridge: 3µm,120Å, 4.6×10mm, 10pcs/pk
Catalog No. NC2052537
Encompass_Preferred
Cartridge: Xtimate C8, 3µm,120Å, 4.6×10mm, pH range: 1.0-12.5, Pore Size: 120Å, USP listing: L7, Carbon Loading: 10%, Surface Area: 320m2/g, endcapped. Matched HPLC Column ID: 3.0-8.0, Matched Guard Column Holder: 00808-01101 or 00808-01108, 10pcs/pk.
Catalog No. NC2052537 Supplier WELCH MATERIALS INC Supplier No. 008080310210
Please call customer service for current pricing. 1-800-766-7000
Cartridge: Xtimate C8, 3µm,120Å, 4.6×10mm, pH range: 1.0-12.5, Pore Size: 120Å, USP listing: L7, Carbon Loading: 10%, Surface Area: 320m2/g, endcapped. Matched HPLC Column ID: 3.0-8.0, Matched Guard Column Holder: 00808-01101 or 00808-01108, 10pcs/pk.