Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corning™ XT Cooling Core for CoolBox™ XT Workstation

Catalog No. 07210081 Shop All Corning Products
Change view
Click to view available options
Color:
Gray
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
07-210-081 Gray
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 07-210-081 Supplier Corning™ Supplier No. 432081
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy

Allows for continuous uninterrupted cooling. Corning™ XT Cooling Core for CoolBox™ XT Workstation provides cooling source workstation when thermo-conductive CoolRack or CoolSink tube and plate module is placed on top.

XT Cooling Core for CoolBox XT, a second one allows for continuous uninterrupted cooling.

TRUSTED_SUSTAINABILITY

Specifications

Color Gray
For Use With (Application) Maintaining sample temperature
For Use With (Equipment) CoolBox XT, CoolBox 2XT, or XT Starter Systems
Quantity 1
Dimensions (L x W x H) 16.2 x 12.1 x 6.1 cm (6.3 x 4.7 x 2.1 in.)
Type XT Cooling Core
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.