Learn More
Bio Rad Laboratories XT MES RUNNING BUFFER 500ML
Catalog No. NC9081971
Encompass
XT MES Running Buffer, 20x, 500mL, Liquid, White, Odorless, Boiling Point: 100degreeC, pH: 6.4 at 20degreeC, Fully miscible in water, No special requirements for storage
Catalog No. NC9081971 Supplier Bio Rad Laboratories Supplier No. 1610789
XT MES Running Buffer, 20x, 500mL, Liquid, White, Odorless, Boiling Point: 100degreeC, pH: 6.4 at 20degreeC, Fully miscible in water, No special requirements for storage