Learn More
Bio Rad Laboratories XT MOPS RUNNING BUFFER 500ML
Catalog No. NC9056724
Encompass
Extended range protein standard, 500ul, 50 applications. Product contains 12 bands from 2kD to 250kD. 250, 150, 100, 50, 37, 20, 15, 10 & 5 kD are blue, 75, 25 and 2kD are pink.
Catalog No. NC9056724 Supplier Bio Rad Laboratories Supplier No. 1610788
Extended range protein standard, 500ul, 50 applications. Product contains 12 bands from 2kD to 250kD. 250, 150, 100, 50, 37, 20, 15, 10 & 5 kD are blue, 75, 25 and 2kD are pink.