Learn More
WELCH MATERIALS INC Xtimate NH2, 5µm, 120Å 4.6×250mm
Catalog No. NC2052236
Encompass_Preferred
Analytical Column, Xtimate NH2, 5μm, 120Å, 4.6×250mm, pH range: 2.0-8.0, Pore Size: 120Å, USP listing: L8, , Surface Area: 450m2/g, not-endcapped.
Catalog No. NC2052236 Supplier WELCH MATERIALS INC Supplier No. 0010321043
Analytical Column, Xtimate NH2, 5μm, 120Å, 4.6×250mm, pH range: 2.0-8.0, Pore Size: 120Å, USP listing: L8, , Surface Area: 450m2/g, not-endcapped.