Learn More
WELCH MATERIALS INC Xtimate Phenyl-Hexyl, 5µm, 10×250mm
Supplier:  WELCH MATERIALS INC 0167021102
Encompass_Preferred
Prep Column, Xtimate Phenyl-Hexyl, 5µm, 10×250mm, pH range: 1.0-12.5, Pore Size: 120Å, USP listing: L11, Carbon Loading: 12%, Surface Area: 320m2/g, endcapped.
Catalog No. NC2052731