Learn More
WELCH MATERIALS INC Xtimate Phenyl-Hexyl, 10µm, 30×250mm
Catalog No. NC2052742
Encompass_Preferred
Prep Column, Xtimate Phenyl-Hexyl, 10µm, 30×250mm, pH range: 1.0-12.5, Pore Size: 120Å, USP listing: L11, Carbon Loading: 12%, Surface Area: 320m2/g, endcapped.
Catalog No. NC2052742 Supplier WELCH MATERIALS INC Supplier No. 0167031108
Add to Cart
Add to Cart
Request a Quote
Prep Column, Xtimate Phenyl-Hexyl, 10µm, 30×250mm, pH range: 1.0-12.5, Pore Size: 120Å, USP listing: L11, Carbon Loading: 12%, Surface Area: 320m2/g, endcapped.