Learn More
Bio Rad Laboratories XT SAMPLE BUFFER, 4X, 10ML
Catalog No. NC9081972
Encompass
XT MOPS running buffer is a denaturing running buffer designed for sharp electrophoretic separations of proteins, particularly those proteins of midsize molecular weight. This running buffer is used with Criterion XT Bis-Tris precast gels.
Catalog No. NC9081972 Supplier Bio Rad Laboratories Supplier No. 1610791
Add to Cart
Add to Cart
XT MOPS running buffer is a denaturing running buffer designed for sharp electrophoretic separations of proteins, particularly those proteins of midsize molecular weight. This running buffer is used with Criterion XT Bis-Tris precast gels.