Learn More
Ankom Technology XT15 Vent Filter
SDP
Supplier:  Ankom Technology X83
Encompass
This part is used to filter solvent vapors and should be replaced every 3 months or after 100 instrument operations. This part is only used in the XT15 and XT15I instruments.
Catalog No. NC1691342