Learn More
COAST Cutlery XT30R Rechargeable Flashlight
Catalog No. 19030064
Change view
Click to view available options
Light Source:
LED
Catalog No. Light Source
19-030-064 LED
1 options
Catalog No. 19-030-064 Supplier COAST Cutlery Supplier No. 31124
Add to Cart
Add to Cart
Go bright all day with high-performance pen lighting. Built to deliver up to 1000 lumens and long run times, the XT30R features five light modes supported by an oversized rechargeable battery.
  • Battery Charging Level Indicator - Check the status of your batteries charge level with our built-in charge level indicator. (Solid red) when charging, (solid green) when fully charged.
  • Built-In USB-C Port - With a built-in USB-C port, your battery can be charged using a USB-C cord. Some COAST models can be charged both inside or outside the light.
  • Dust Resistant - Take on any dirty work environment with this dust resistant light.
  • Pro-Tek Charging - Charge directly from AC, DC, and USB. Flashlights and headlamps with COAST PRO-TEK Charging include a direct-connect port for powering back up.
  • Pure Beam Focusing Optic - Pack two powerful beams into one and seamlessly transition - with no halos or hot spots - between short and long-range viewing. COAST's patented focusing technology adapts to your need. The Ultra View Flood beam delivers edge-to-edge clarity in a broad circle, and the BULLS-EYE™ Spot Beam sends a sharp ray of light at a distance with a transitional halo around the spot to increase the effective viewing area.
  • Twist Focus - Dial in the shape of light you need for the situation. COAST's TWIST FOCUS™ system makes it easy to move between spot and flood beams by simply rotating the head of the flashlight.
  • The XT30R features five light modes - medium, high, Turbo, low, and MoonGlow
Specifications
Light Source LED
Voltage 3.7 V
Color Black
Product Type Rechargeable LED Flashlight