Learn More
Mitegen JBScreen Kinase HTS L Crystallization Screen, 1.7mL/Well, For High-Throughput Kinase Protein Crystallization
SDP
Catalog No. 501187263
Encompass
The JBScreen Kinase HTS L Crystallization Screen offers a highly specialized and efficient solution for high-throughput crystallization screening of kinase proteins. Each well contains 1.7 ml of solution, providing ample volume for setting up multiple crystallization trials tailored to the unique requirements of kinase targets. Designed to streamline your screening workflow, this dedicated screen assists researchers in quickly identifying promising conditions for kinase crystal growth, minimizing extensive trial-and-error. Its optimized formulation contributes to improving crystal quality and reproducibility, essential for subsequent X-ray diffraction analysis and accurate structural determination. This specialized screen is an indispensable tool for laboratories engaged in structural biology and protein science focusing on kinases, significantly enhancing the efficiency and success rate of their crystallization pipeline for challenging targets.
Catalog No. 50-118-7263 Supplier Mitegen Supplier No. MCS204L
The JBScreen Kinase HTS L Crystallization Screen offers a highly specialized and efficient solution for high-throughput crystallization screening of kinase proteins. Each well contains 1.7 ml of solution, providing ample volume for setting up multiple crystallization trials tailored to the unique requirements of kinase targets. Designed to streamline your screening workflow, this dedicated screen assists researchers in quickly identifying promising conditions for kinase crystal growth, minimizing extensive trial-and-error. Its optimized formulation contributes to improving crystal quality and reproducibility, essential for subsequent X-ray diffraction analysis and accurate structural determination. This specialized screen is an indispensable tool for laboratories engaged in structural biology and protein science focusing on kinases, significantly enhancing the efficiency and success rate of their crystallization pipeline for challenging targets.