Learn More
Waters Corp Sentry Guard Cartridge, XTerra MS C18, 125Å, 3.5 μm, 2.1 mm x 20 mm
Catalog No. 50819071 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This XTerra MS C18 Sentry Guard Cartridge is designed for use in mass spectrometry applications, featuring a hybrid-based, reversed-phase C18 material for robust and efficient performance.
  • Hybrid-based C18 reversed-phase material.
  • 125Å pore size.
  • 3.5μm particle size.
  • 2.1 mm ID and 20 mm length.
  • Pack of 2 cartridges.
  • Guard cartridge type.
  • USP L1 phase type.
Catalog No. 50-819-071 Supplier Waters Corp Supplier No. 186000636
This XTerra MS C18 Sentry Guard Cartridge is designed for use in mass spectrometry applications, featuring a hybrid-based, reversed-phase C18 material for robust and efficient performance.
  • Hybrid-based C18 reversed-phase material.
  • 125Å pore size.
  • 3.5μm particle size.
  • 2.1 mm ID and 20 mm length.
  • Pack of 2 cartridges.
  • Guard cartridge type.
  • USP L1 phase type.