Learn More
Waters Corp XTerra Column, C18, 125 Å, 3.5 μm, 3 mm × 150 mm
Catalog No. 502392949 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The XTerra MS C18 Column features an electronic eConnect column tag for automatic identification, tracking, and digital tracing of column use history. These NFC-enabled columns are compatible with any HPLC system, with enhanced advantages when used with the Alliance iS System running Empower software.
  • Equipped with an electronic eConnect column tag containing unique column-specific information.
  • NFC-enabled device-tagged columns developed to automatically identify, track, and digitally trace column use history.
  • Can be used on any HPLC.
  • Advantages are unlocked when used with the Alliance iS System running Empower software.
Catalog No. 50-239-2949 Supplier Waters Corp Supplier No. 186000422RF
Add to Cart
Add to Cart
The XTerra MS C18 Column features an electronic eConnect column tag for automatic identification, tracking, and digital tracing of column use history. These NFC-enabled columns are compatible with any HPLC system, with enhanced advantages when used with the Alliance iS System running Empower software.
  • Equipped with an electronic eConnect column tag containing unique column-specific information.
  • NFC-enabled device-tagged columns developed to automatically identify, track, and digitally trace column use history.
  • Can be used on any HPLC.
  • Advantages are unlocked when used with the Alliance iS System running Empower software.