Learn More
Waters Corp XTerra MS C18 Column, 5 μm, 3 × 100 mm
Catalog No. 502390692 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
XTerra MS C18 columns are general-purpose, hybrid-based, reversed-phase C18 columns designed for compatibility with mass spectrometry applications. These first-generation columns provide superior pH stability compared to a silica-based column. They are also equipped with an electronic eConnect column tag for enhanced tracking and tracing capabilities.
  • Compatible with mass spectrometry applications.
  • Offers superior pH stability compared to silica-based columns.
  • Equipped with an electronic eConnect column tag for automatic identification, tracking, and digital tracing of column use history.
  • NFC-enabled device-tagged.
  • Can be used on any HPLC system.
  • Advantages are unlocked when used with compatible systems running Empower software.
Catalog No. 50-239-0692 Supplier Waters Corp Supplier No. 186000466RF
XTerra MS C18 columns are general-purpose, hybrid-based, reversed-phase C18 columns designed for compatibility with mass spectrometry applications. These first-generation columns provide superior pH stability compared to a silica-based column. They are also equipped with an electronic eConnect column tag for enhanced tracking and tracing capabilities.
  • Compatible with mass spectrometry applications.
  • Offers superior pH stability compared to silica-based columns.
  • Equipped with an electronic eConnect column tag for automatic identification, tracking, and digital tracing of column use history.
  • NFC-enabled device-tagged.
  • Can be used on any HPLC system.
  • Advantages are unlocked when used with compatible systems running Empower software.