Learn More
Waters Corp XTerra MS C18 Column, 125Å, 3.5 μm, 4.6 mm × 50 mm
Catalog No. 502391517 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
XTerra MS C18 columns are hybrid-based, reversed-phase C18 columns for mass spectrometry. They offer superior pH stability (1 to 12) and operate up to 6000 psi (415 Bar), with an eConnect Column Tag.
  • Designed for mass spectrometry with superior pH stability (1 to 12) and high pressure (up to 6000 psi / 415 Bar) resistance.
  • Features a second-generation eConnect column tag, providing product-specific identification and NFC access to support.
  • Enables automatic data import/searchability via Empower CDS, simplifying setup and reducing manual data entry.
  • The tamper-proof tag enhances compliance, auto-detectability, and prevents interference in temperature-sensitive LC methods.
  • Performance is equivalent to untagged counterparts in manufacturing, quality, and function.
Catalog No. 50-239-1517 Supplier Waters Corp Supplier No. 186000432RF
XTerra MS C18 columns are hybrid-based, reversed-phase C18 columns for mass spectrometry. They offer superior pH stability (1 to 12) and operate up to 6000 psi (415 Bar), with an eConnect Column Tag.
  • Designed for mass spectrometry with superior pH stability (1 to 12) and high pressure (up to 6000 psi / 415 Bar) resistance.
  • Features a second-generation eConnect column tag, providing product-specific identification and NFC access to support.
  • Enables automatic data import/searchability via Empower CDS, simplifying setup and reducing manual data entry.
  • The tamper-proof tag enhances compliance, auto-detectability, and prevents interference in temperature-sensitive LC methods.
  • Performance is equivalent to untagged counterparts in manufacturing, quality, and function.