Learn More
Waters Corp Waters XTerra MS C18 Column, 2.5 μm, 4.6 mm, 75 mm, 125 Å
Catalog No. 50780520 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
XTerra MS C18 columns are general-purpose, hybrid-based, reversed-phase C18 columns designed for mass spectrometry applications. These first-generation columns offer superior pH stability compared to silica-based columns. They are constructed using Hybrid Particle Technology for excellent separation efficiency and improved pH stability.
  • General purpose, hybrid-based, reversed-phase C18 columns.
  • Designed for mass spectrometry applications.
  • Superior pH stability compared to silica-based columns.
  • Offers sharp peak forms, high sensitivity, and large peak capacities.
  • Utilizes trifunctional bonding chemistry for long lifetimes over a broad pH range.
  • Ensures maximum throughput, superb resolution, and ultra-low column bleed.
  • Individually tested with certificates of batch analysis and column efficiency.
Catalog No. 50-780-520 Supplier Waters Corp Supplier No. 186000981
Add to Cart
Add to Cart
XTerra MS C18 columns are general-purpose, hybrid-based, reversed-phase C18 columns designed for mass spectrometry applications. These first-generation columns offer superior pH stability compared to silica-based columns. They are constructed using Hybrid Particle Technology for excellent separation efficiency and improved pH stability.
  • General purpose, hybrid-based, reversed-phase C18 columns.
  • Designed for mass spectrometry applications.
  • Superior pH stability compared to silica-based columns.
  • Offers sharp peak forms, high sensitivity, and large peak capacities.
  • Utilizes trifunctional bonding chemistry for long lifetimes over a broad pH range.
  • Ensures maximum throughput, superb resolution, and ultra-low column bleed.
  • Individually tested with certificates of batch analysis and column efficiency.