Learn More
Waters Corp Waters Corporation XTerra MS C18 Column, 3.5 μm, 125 Å, 3 mm × 30 mm
Catalog No. 50780231 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
XTerra MS C18 columns are designed for mass spectrometry applications. These hybrid-based, reversed-phase C18 columns offer superior pH stability compared to silica-based columns.
  • General purpose, hybrid-based, reversed-phase C18 columns.
  • Compatible with mass spectrometry applications.
  • Superior pH stability compared to silica-based columns.
Catalog No. 50-780-231 Supplier Waters Corp Supplier No. 186000412
Add to Cart
Add to Cart
XTerra MS C18 columns are designed for mass spectrometry applications. These hybrid-based, reversed-phase C18 columns offer superior pH stability compared to silica-based columns.
  • General purpose, hybrid-based, reversed-phase C18 columns.
  • Compatible with mass spectrometry applications.
  • Superior pH stability compared to silica-based columns.