Learn More
Waters Corp Waters Corporation Column, C18, 3.5 μm, 3 mm ID, 50 mm Length
Supplier:  Waters Corp 186000414
Encompass_Preferred
  • Designed for mass spectrometry (MS) applications.
  • Features crisp peak forms, high sensitivity, and large peak capacities.
  • Offers long column life over a wide pH range (1 to 12) due to tri-functional bonding chemistry.
  • Provides maximum throughput with exceptional resolution and minimal column bleed.
  • Uses hybrid particle technology (inorganic silica and organic organosiloxane components).
  • Provides superior separation efficiency and enhanced pH stability.
Catalog No. 50-780-233