Learn More
Waters Corp Waters Corporation HPLC Column, C18, 3.5 μm, 4.6 mm ID, 250 mm Length, 125 Å
Catalog No. 50780699 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This HPLC column is a general-purpose, hybrid-based, reversed-phase C18 column, specifically designed for compatibility with mass spectrometry applications. It offers superior pH stability compared to traditional silica-based columns.
  • General-purpose, hybrid-based, reversed-phase C18 column.
  • Designed for compatibility with mass spectrometry (MS) applications.
  • Provides superior pH stability compared to silica-based columns.
  • Suitable for use with both aqueous and organic mobile phases.
  • Well-suited for the separation of small to medium-sized polar and non-polar compounds.
  • Hybrid particle technology ensures excellent peak shapes for acidic and basic compounds.
  • Minimizes secondary interactions, resulting in sharp peak shapes.
  • Offers robust performance in simple and complex matrices.
Catalog No. 50-780-699 Supplier Waters Corp Supplier No. 186001470
This HPLC column is a general-purpose, hybrid-based, reversed-phase C18 column, specifically designed for compatibility with mass spectrometry applications. It offers superior pH stability compared to traditional silica-based columns.
  • General-purpose, hybrid-based, reversed-phase C18 column.
  • Designed for compatibility with mass spectrometry (MS) applications.
  • Provides superior pH stability compared to silica-based columns.
  • Suitable for use with both aqueous and organic mobile phases.
  • Well-suited for the separation of small to medium-sized polar and non-polar compounds.
  • Hybrid particle technology ensures excellent peak shapes for acidic and basic compounds.
  • Minimizes secondary interactions, resulting in sharp peak shapes.
  • Offers robust performance in simple and complex matrices.