Learn More
Waters Corp Waters Corporation XTerra MS C18 Column, 3.5 μm, 3.0 mm x 150 mm
Supplier:  Waters Corp 186000422
Encompass_Preferred
The XTerra HPLC Column is a first-generation hybrid particle stationary phase, combining silica's efficiency with polymer stability. These methylene bridged particles are designed for Mass Spectrometry applications.
  • High efficiency separations.
  • Expanded pH range (1–12) compared to silica-based particles.
  • Rugged material with high mechanical strength.
  • Excellent peak shape for bases, with good sensitivity and large peak capacities.
  • Trifunctional bonding chemistries deliver long lifetimes over a wide pH range.
  • Offers high throughput, excellent resolution, and ultra-low bleed.
  • Homogenous particle coverage with less silanols for superior peak shape.
  • Easy scale-up from analytical to preparative chromatography.
  • Increased mass loading (up to 60x) at high pH.
Catalog No. 50-818-707