Learn More
Waters Corp Waters Column, C18, 125Å, 3.5 μm, 4.6 mm × 50 mm
Supplier:  Waters Corp 186000432
Encompass_Preferred
These general purpose, hybrid-based, reversed-phase C18 columns are designed to be compatible with mass spectrometry applications, offering superior pH stability compared to silica-based columns.
  • General purpose, hybrid-based, reversed-phase C18 columns.
  • Designed to be compatible with mass spectrometry applications.
  • Provides superior pH stability compared to silica-based columns.
  • Remarkable pH range spanning from 1 to 12, accommodating a wide spectrum of acidic and basic conditions.
  • Exceptional durability, withstanding high pressures up to 6000 psi (415 Bar).
  • Robust capabilities for various chromatographic applications, ensuring reliable and efficient separations across a range of experimental conditions.
Catalog No. 50-819-147