Learn More
Waters Corp Waters XTerra MS C18 Column, 125Å, 5 μm, 3 mm X 150 mm
Supplier:  Waters Corp 186000470
Encompass_Preferred
XTerra MS C18 columns are general-purpose, hybrid-based, reversed-phase C18 columns compatible with mass spectrometry. They offer superior pH stability compared to silica-based columns, with an innovative hybrid particle substrate for exceptional peak shape and consistent results. Reduced silanol activity minimizes contamination. A wide pH range of 1 to 12 accommodates diverse analytical needs.
  • Designed to be compatible with mass spectrometry applications
  • Provides superior pH stability compared to silica-based columns
  • Innovative hybrid particle substrate for unparalleled pH stability and exceptional peak shape
  • Reduced silanol activity minimizes the risk of sample contamination
  • Wide pH range spanning from 1 to 12
  • Ensures accurate and reproducible results
  • Thoughtfully end-capped to prevent sample loss
Catalog No. 50-780-287