Learn More
Waters Corp Waters XTerra MS C18 Column, 5 μm, 3 mm x 50 mm
Supplier:  Waters Corp 186000462
Encompass_Preferred
The Waters XTerra MS C18 Column is a general-purpose, hybrid-based, reversed-phase C18 column engineered for mass spectrometry applications. It delivers superior pH stability and high efficiency, ensuring sharp peak forms and large peak capacities for optimal analytical performance.
  • General purpose, hybrid-based, reversed-phase C18 columns.
  • Compatible with mass spectrometry applications.
  • Provides superior pH stability compared to silica-based columns.
  • Features sharp peak forms, good sensitivity, and large peak capacities for mass spectrometry applications.
  • Utilizes tri-functional bonding chemistry for long lifetime across a wide pH range.
  • Offers exceptional resolution, extremely low column bleed, and maximum throughput.
  • Manufactured with controlled cleanliness and chemical composition using ultrapure reagents.
Catalog No. 50-780-279