Learn More
Waters Corp Waters XTerra MS C18 Column, 5 μm, 4.6 x 30 mm
Catalog No. 50780514 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
XTerra HPLC Columns are hybrid particle (silica co-polymer) stationary phases. Their methylene bridged particles combine high efficiency with polymer stability. Designed for Mass Spectrometry (MS) applications, these columns provide sharp peaks, good sensitivity, large peak capacities, and an expanded pH range.
  • Hybrid (silica co-polymer) stationary phase.
  • Methylene bridged particles for efficiency and polymer stability.
  • Expanded pH range (1–12) for versatile separations.
  • High mechanical strength and excellent peak shape for bases.
  • Optimized for Mass Spectrometry (MS) applications.
  • Sharp peaks, good sensitivity, and large peak capacity.
  • Trifunctional bonding chemistries ensure long column lifetimes.
  • High throughput, excellent resolution, and ultra-low bleed.
  • Easy scale-up from analytical to preparative chromatography.
Catalog No. 50-780-514 Supplier Waters Corp Supplier No. 186000878
Add to Cart
Add to Cart
XTerra HPLC Columns are hybrid particle (silica co-polymer) stationary phases. Their methylene bridged particles combine high efficiency with polymer stability. Designed for Mass Spectrometry (MS) applications, these columns provide sharp peaks, good sensitivity, large peak capacities, and an expanded pH range.
  • Hybrid (silica co-polymer) stationary phase.
  • Methylene bridged particles for efficiency and polymer stability.
  • Expanded pH range (1–12) for versatile separations.
  • High mechanical strength and excellent peak shape for bases.
  • Optimized for Mass Spectrometry (MS) applications.
  • Sharp peaks, good sensitivity, and large peak capacity.
  • Trifunctional bonding chemistries ensure long column lifetimes.
  • High throughput, excellent resolution, and ultra-low bleed.
  • Easy scale-up from analytical to preparative chromatography.