Learn More
Waters Corp Waters Column, C18, 5 μm, 4.6 mm X 100 mm, 125 Å
Catalog No. 50780303 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
XTerra MS C18 columns are general-purpose, hybrid-based, reversed-phase C18 columns designed to be compatible with mass spectrometry applications. These first-generation columns offer superior pH stability compared to silica-based columns.
  • Tri-functional bonding chemistry offers long life.
  • Stable over a pH range of 1–12.
  • High throughput, excellent resolution, and minimal column bleed.
  • Hybrid particle technology combines inorganic (silica) and organic (organosiloxane) components.
  • Provides great separation efficiency and better pH stability compared to silica-based reversed-phase packing materials.
  • Manufactured using ultrapure reagents for controlled chemical composition and purity.
  • Meticulous monitoring and control at every production stage.
  • Each column is individually examined for quality assurance.
Catalog No. 50-780-303 Supplier Waters Corp Supplier No. 186000486
XTerra MS C18 columns are general-purpose, hybrid-based, reversed-phase C18 columns designed to be compatible with mass spectrometry applications. These first-generation columns offer superior pH stability compared to silica-based columns.
  • Tri-functional bonding chemistry offers long life.
  • Stable over a pH range of 1–12.
  • High throughput, excellent resolution, and minimal column bleed.
  • Hybrid particle technology combines inorganic (silica) and organic (organosiloxane) components.
  • Provides great separation efficiency and better pH stability compared to silica-based reversed-phase packing materials.
  • Manufactured using ultrapure reagents for controlled chemical composition and purity.
  • Meticulous monitoring and control at every production stage.
  • Each column is individually examined for quality assurance.