Learn More
Waters Corp Waters XTerra MS C18 Column, 5 μm, 125 Å, 3.9 mm × 150 mm
Supplier:  Waters Corp 186000478
Encompass_Preferred
XTerra MS C18 columns are general-purpose, hybrid-based, reversed-phase C18 columns designed for mass spectrometry applications. They provide superior pH stability compared to silica-based columns, offering sharp peaks, good sensitivity, and large peak capacities. Trifunctional bonding chemistry ensures extended lifetime over a wide pH range (1–12).
  • Designed for mass spectrometry applications, delivering sharp peaks, good sensitivity, and large peak capacities.
  • Trifunctional bonding chemistry provides long lifetimes over a wide pH range (1–12) with excellent resolution and ultra-low bleed.
  • Hybrid particle technology offers high mechanical strength, efficiency, and excellent peak shape.
  • Facilitates analytical to preparative scale-up.
  • Proprietary end-capping for superior peak shape.
Catalog No. 50-818-711