Learn More
Waters Corp Waters Column, C18, 5 μm, 125 Å, 4.6 mm, 250 mm
Catalog No. 50818712 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These hybrid-based, reversed-phase C18 columns are designed for mass spectrometry, offering superior pH stability and mechanical strength compared to silica-based columns. They provide high efficiency, superior peak shape for bases, and are suitable for easy scale-up.
  • Superior pH stability and high mechanical strength.
  • High efficiency and superior peak shape for bases.
  • Easy scale-up for analytical and preparative chromatography.
  • Optimized for mass spectrometry with sharp peaks, good sensitivity, and large peak capacities.
  • Tri-functional bonding ensures long lifetimes across a wide pH range (1 to 12).
  • Delivers maximum throughput, excellent resolution, and ultra-low bleed.
  • Manufactured with ultrapure reagents, individually tested under narrow tolerances.
  • Produced in cGMP, ISO 9002 certified facilities for superior reproducibility.
Catalog No. 50-818-712 Supplier Waters Corp Supplier No. 186000494
These hybrid-based, reversed-phase C18 columns are designed for mass spectrometry, offering superior pH stability and mechanical strength compared to silica-based columns. They provide high efficiency, superior peak shape for bases, and are suitable for easy scale-up.
  • Superior pH stability and high mechanical strength.
  • High efficiency and superior peak shape for bases.
  • Easy scale-up for analytical and preparative chromatography.
  • Optimized for mass spectrometry with sharp peaks, good sensitivity, and large peak capacities.
  • Tri-functional bonding ensures long lifetimes across a wide pH range (1 to 12).
  • Delivers maximum throughput, excellent resolution, and ultra-low bleed.
  • Manufactured with ultrapure reagents, individually tested under narrow tolerances.
  • Produced in cGMP, ISO 9002 certified facilities for superior reproducibility.