Learn More
Waters Corp Waters Column, XTerra MS C18, 125Å, 5 μm, 4.6 mm X 50 mm
Supplier:  Waters Corp 186000482
Encompass_Preferred
XTerra MS C18 columns are hybrid-based, reversed-phase C18 columns designed for mass spectrometry applications, offering superior pH stability and delivering sharp peak forms, exceptional sensitivity, and large peak capacities.
  • Superior pH stability.
  • Designed for mass spectrometry, with sharp peak forms and high sensitivity.
  • Large peak capacities.
  • Tri-functional bonding chemistry for long life (wide pH range).
  • High throughput, resolution, and minimal column leakage.
  • Ultrapure reagents for controlled composition and purity.
  • Individually tested; batch analysis and efficiency certificates provided.
  • Hybrid particle technology (organic/inorganic benefits).
  • Functions across 1–12 pH range.
  • Max pressure: 6,000 psi (415 Bar).
Catalog No. 50-818-626