Learn More
Waters Corp XTerra Hybrid MS C18 Method Validation Kit, 125Å, 3.5 µm, 4.6 mm X 150 mm, 3/pk
Supplier:  Waters Corp 186000826
Encompass_Preferred
Method Validation Kits allow the chromatographer to check a method for long-term ruggedness. The kit consists of three XTerra columns packed with three different batches. Kits are available in a variety of particle sizes and dimensions.
Catalog No. 50-780-490