Learn More
Waters Corp Waters Sentry Guard Cartridge, 125 Å, 3.5 μm, 3.9 mm × 20 mm
Catalog No. 50780433 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This product is a general-purpose, hybrid-based, reversed-phase C18 column designed for compatibility with mass spectrometry applications. It offers superior pH stability compared to traditional silica-based columns.
  • Compatible with mass spectrometry applications.
  • Offers superior pH stability (pH range of 1 to 12).
  • Features C18 chemistry, reversed-phase separation, and a hybrid particle substrate.
  • Withstands a maximum pressure of 6000 psi (415 Bar).
  • Endcapped, with MS C18 bonding technology and low silanol activity.
  • Spherical particle shape, 3.5 μm particle size, and 125 Å pore size.
  • Functions as a guard cartridge with a surface area of 175.
  • Suitable for HPLC systems, meeting USP classification L1.
  • Dimensions: 3.9 mm inner diameter, 20 mm length, and 16% carbon load.
Catalog No. 50-780-433 Supplier Waters Corp Supplier No. 186000644
This product is a general-purpose, hybrid-based, reversed-phase C18 column designed for compatibility with mass spectrometry applications. It offers superior pH stability compared to traditional silica-based columns.
  • Compatible with mass spectrometry applications.
  • Offers superior pH stability (pH range of 1 to 12).
  • Features C18 chemistry, reversed-phase separation, and a hybrid particle substrate.
  • Withstands a maximum pressure of 6000 psi (415 Bar).
  • Endcapped, with MS C18 bonding technology and low silanol activity.
  • Spherical particle shape, 3.5 μm particle size, and 125 Å pore size.
  • Functions as a guard cartridge with a surface area of 175.
  • Suitable for HPLC systems, meeting USP classification L1.
  • Dimensions: 3.9 mm inner diameter, 20 mm length, and 16% carbon load.