Learn More
Waters Corp Waters Sentry Guard Cartridge, C18, 5 μm, 3 mm x 20 mm
Catalog No. 50780445 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These MS C18 columns are general purpose, hybrid-based, reversed-phase columns designed to be compatible with mass spectrometry applications. These first-generation columns offer superior pH stability compared to silica-based columns.
  • C18 chemistry
  • Reversed phase separation mode
  • Hybrid particle substrate
  • pH range of 1 to 12
  • Maximum pressure of 6000 psi (415 Bar)
  • Endcapped
  • MS C18 bonding technology
  • Low silanol activity
  • Spherical particle shape
  • 5 μm particle size
  • 125 Å pore size
  • Sentry guard cartridge format
  • 175 surface area
  • HPLC system compatible
  • USP classification L1
  • 3 mm inner diameter
  • 20 mm length
  • 16% carbon load
  • 2 units per package
Catalog No. 50-780-445 Supplier Waters Corp Supplier No. 186000656
These MS C18 columns are general purpose, hybrid-based, reversed-phase columns designed to be compatible with mass spectrometry applications. These first-generation columns offer superior pH stability compared to silica-based columns.
  • C18 chemistry
  • Reversed phase separation mode
  • Hybrid particle substrate
  • pH range of 1 to 12
  • Maximum pressure of 6000 psi (415 Bar)
  • Endcapped
  • MS C18 bonding technology
  • Low silanol activity
  • Spherical particle shape
  • 5 μm particle size
  • 125 Å pore size
  • Sentry guard cartridge format
  • 175 surface area
  • HPLC system compatible
  • USP classification L1
  • 3 mm inner diameter
  • 20 mm length
  • 16% carbon load
  • 2 units per package