Learn More
Waters Corp OBD Prep Column, C18, 10 μm, 125 Å, 10 mm x 150 mm
Catalog No. 502391265 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
XTerra Prep columns utilize Hybrid Particle Technology for isolation and purification in discovery and development laboratories. Engineered for preparative applications, these columns operate at pH 1–12, enabling 60X more material loading. This increased capacity allows fewer injections for the same pure material or more material per injection. For discovery, this means shorter columns and faster material production.
  • Utilizes Hybrid Particle Technology for isolation and purification
  • Engineered specifically for preparative applications
  • Operates routinely at pH 1–12
  • Enables loading up to 60X more material onto the column
  • Increased capacity results in fewer injections for the same amount of pure material or more material with the same number of injections
  • Greater capacity allows use of a shorter column for faster material production
Catalog No. 50-239-1265 Supplier Waters Corp Supplier No. 186008129
XTerra Prep columns utilize Hybrid Particle Technology for isolation and purification in discovery and development laboratories. Engineered for preparative applications, these columns operate at pH 1–12, enabling 60X more material loading. This increased capacity allows fewer injections for the same pure material or more material per injection. For discovery, this means shorter columns and faster material production.
  • Utilizes Hybrid Particle Technology for isolation and purification
  • Engineered specifically for preparative applications
  • Operates routinely at pH 1–12
  • Enables loading up to 60X more material onto the column
  • Increased capacity results in fewer injections for the same amount of pure material or more material with the same number of injections
  • Greater capacity allows use of a shorter column for faster material production